Order Entry
Export
ContactUsLinkComponent
 
Anti-p53 Rabbit Recombinant Antibody [clone: TP53/7002R]
  ANTIA278661-100
New Product
undefined
Anti-p53 Rabbit Recombinant Antibody [clone: TP53/7002R]
  ANTIA278661-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    Antigen NY-CO-13
  • Antigen symbol:
    p53
  • Clonality:
    Recombinant
  • Clone:
    TP53/7002R
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P04637
  • Antigen synonyms:
    P53_HUMAN|Tp53|BCC7|Cellular tumor antigen p53|Mutant tumor protein 53|LFS1|Phosphoprotein p53|FLJ92943|p53 tumor suppressor
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
  • Molecular weight:
    53 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full length human TP53 protein.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Recombinant rabbit monoclonal [TP53/7002R] antibody to p53 for Flow cytometry and IHC-P with samples derived from human.

  • Validated applications: Flow cytometry, IHC-P
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: p53
Clonality: Recombinant
Clone: TP53/7002R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human