Anti-StAR Rabbit Recombinant Antibody [clone: STAR/3915R]
ANTIA278655-100
New Product
- Antibody type:Primary
- Antigen name:Cholesterol trafficker
- Antigen symbol:StAR
- Clonality:Recombinant
- Clone:STAR/3915R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P49675
- Antigen synonyms:START domain containing 1|StAR|StARD1|mitochondrial|Luteinizing hormone induced protein|START domain-containing protein 1|START domain containing protein 1|StAR related lipid transfer (START) domain containing 1|Mitochondrial steroid acute regulatory protein
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:30 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 39 - 108, of human StAR protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Recombinant rabbit monoclonal [STAR/3915R] antibody to StAR for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: StAR
Clonality: Recombinant
Clone: STAR/3915R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human