Anti-CD45RA Rabbit Recombinant Antibody [clone: PTPRC/7018R]
Catalog # ANTIA278641-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:B220
- Antigen symbol:CD45RA
- Clonality:Recombinant
- Clone:PTPRC/7018R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P08575
- Antigen synonyms:CD45R|CD45|CD45RA|GP180|Leukocyte common antigen|L-CA|LCA|CD45 antigen|EC 3.1.3.48
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:180 - 220 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full length human PTPRC protein.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [PTPRC/7018R] antibody to CD45RA for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC-P: 0,5 to 1 µg/ml
Type: Primary
Antigen: CD45RA
Clonality: Recombinant
Clone: PTPRC/7018R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1 kappa
Reactivity: Human