Order Entry
Export
ContactUsLinkComponent
Anti-Prolactin Rabbit Recombinant Antibody [clone: PRL/4908R]
Anti-Prolactin Rabbit Recombinant Antibody [clone: PRL/4908R]
Catalog # ANTIA278639-100
Supplier:  Antibodies.com
New Product
undefined
Anti-Prolactin Rabbit Recombinant Antibody [clone: PRL/4908R]
Catalog # ANTIA278639-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    AV290867
  • Antigen symbol:
    Prolactin
  • Clonality:
    Recombinant
  • Clone:
    PRL/4908R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P01236
  • Antigen synonyms:
    Growth hormone A1|Mammotropin|Prl1a1|GHA1|Decidual prolactin|PRL|Luteotropic Hormone |OTTMUSP00000000651|Lactogenic Hormone
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
  • Molecular weight:
    27 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment, around amino acids 63 - 201, of human Prolactin protein. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [PRL/4908R] antibody to Prolactin for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Prolactin
Clonality: Recombinant
Clone: PRL/4908R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human