Anti-Cytokeratin 18 Rabbit Monoclonal Antibody [Clone: ARC2303]
ANTIA305288-100
New Product
- Antibody type:Primary
- Antigen name:Cell proliferation inducing gene 46 protein
- Antigen symbol:Cytokeratin 18
- Clonality:Monoclonal
- Clone:ARC2303
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P05783
- Antigen synonyms:CK18|Cell proliferation inducing protein 46|Cytokeratin 18|CYK18|Cytokeratin endo B|Cell proliferation-inducing gene 46 protein|CK 18|CK-18|CYK 18
- Storage buffer:Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,02% sodium azide
- Molecular weight:48 kDa
- Sequence:ADIRAQYDELARKNREELDKYWSQQIEESTTVVTTQSAEVGAAETTLTELRRTVQSLEIDLDSMRNLKASLENSLREVEARYALQMEQLNGILLHLESELAQTRAEGQRQAQEYEALLNIKVKLEAEIATYRRLLEDGEDFNLGDALDSSNSMQTIQKTTTRRIVDGKVVSETNDTKVLRH
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 250 - 430 of human Cytokeratin 18 (KRT18) (P05783).
- Tested applications:IHC, ICC/IF
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Rabbit monoclonal [ARC2303] antibody to Cytokeratin 18 for WB, IHC and ICC/IF with samples derived from human, mouse and rat.
- Validated applications: WB, IHC, ICC/IF
- Recommended dilutions: WB: 1:1000 to 1:3000, IHC: 1:50 to 1:200, ICC/IF: 1:50 to 1:200
Type: Primary
Antigen: Cytokeratin 18
Clonality: Monoclonal
Clone: ARC2303
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat