Order Entry
Export
ContactUsLinkComponent
 
Anti-SLC38A1/NAT2 Mouse Monoclonal Antibody [Clone: S104-32]
Catalog # ANTIA305004-100
Supplier:  Antibodies.com
New Product
undefined
Anti-SLC38A1/NAT2 Mouse Monoclonal Antibody [Clone: S104-32]
Catalog # ANTIA305004-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    SLC38A1 / NAT2
  • Clonality:
    Monoclonal
  • Clone:
    S104-32
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q9JM15
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,4, with 50% glycerol and 0,09% sodium azide
  • Molecular weight:
    ~50 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Fusion protein amino acids 1 - 63 of rat SNAT1.
  • Tested applications:
    IHC, ICC/IF
  • Purification:
    Protein G purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [S104-32] antibody to SLC38A1/NAT2 for WB, IHC and ICC/IF with samples derived from human, mouse and rat.

  • Validated applications: WB, IHC, ICC/IF
  • Recommended dilutions: WB: 1:1000

Type: Primary
Antigen: SLC38A1/NAT2
Clonality: Monoclonal
Clone: S104-32
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat