Order Entry
Export
ContactUsLinkComponent
Anti-LRP4 Mouse Monoclonal Antibody [Clone: S207-27]
Anti-LRP4 Mouse Monoclonal Antibody [Clone: S207-27]
Catalog # ANTIA304980-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-LRP4 Mouse Monoclonal Antibody [Clone: S207-27]
Catalog # ANTIA304980-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    KIAA0816
  • Antigen symbol:
    LRP4
  • Clonality:
    Monoclonal
  • Clone:
    S207-27
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q8VI56
  • Antigen synonyms:
    LDLR dan|LRP-4|Low-density lipoprotein receptor-related protein 4|MEGF7|Low density lipoprotein receptor related protein 4|Multiple epidermal growth factor like domains 7|LRP10|LRP4|LRP4_HUMAN
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,4, with 50% glycerol and 0,09% sodium azide
  • Molecular weight:
    ~215 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Fusion protein amino acids 26 - 350 (extracellular N - terminus) of mouse LTP4 .
  • Tested applications:
    ICC/IF
  • Purification:
    Protein G purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [S207-27] antibody to LRP4 for WB and ICC/IF with samples derived from human, mouse and rat.

  • Validated applications: WB, ICC/IF
  • Recommended dilutions: WB: 1:1000, ICC/IF: 1:100

Type: Primary
Antigen: LRP4
Clonality: Monoclonal
Clone: S207-27
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat