Anti-NrCAM Mouse Monoclonal Antibody [Clone: S364-51]
ANTIA304970-100
New Product
- Antibody type:Primary
- Antigen name:Bravo
- Antigen symbol:NrCAM
- Clonality:Monoclonal
- Clone:S364-51
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q810U4
- Antigen synonyms:Nr CAM|Ng CAM related|Nr-CAM|NgCAM related cell adhesion molecule|Neuronal surface protein Bravo|Ng-CAM-related|hBravo|NgCAM-related cell adhesion molecule|Neuronal cell adhesion molecule
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 50% glycerol and 0,1% sodium azidelak
- Molecular weight:~160 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Fusion protein amino acids 30 - 845 (extracellular domain) of mouse NrCAM. Rat: 96% identity (795/822 amino acids identical). Human: 91% identity (753/822 amino acids identical) ~50% identity with Neurofascin..
- Tested applications:ICC/IF
- Purification:Protein G purification
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [S364-51] antibody to NrCAM for WB and ICC/IF with samples derived from human, mouse and rat.
- Validated applications: WB, ICC/IF
- Recommended dilutions: WB: 1:1000, ICC/IF: 1:100
Type: Primary
Antigen: NrCAM
Clonality: Monoclonal
Clone: S364-51
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat