Anti-TRP 7 Mouse Monoclonal Antibody [Clone: N64A/36]
Catalog # ANTIA304950-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Capacitative calcium channel TRPC7
- Antigen symbol:TRP 7
- Clonality:Monoclonal
- Clone:N64A/36
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q9HCX4
- Antigen synonyms:hTRP7|mTRP7|Likley ortholog of mouse transient receptor potential cation channel subfamily C member 7|KNP3|Putative capacitative calcium channel|cation channel, subfamily C, member 7|transient receptor potential-related channel 7, a novel putative Ca2+ channel protein|Transient receptor protein 7|Short transient receptor potential channel 7
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 50% glycerol and 0,09% sodium azide
- Molecular weight:~100 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide amino acids 845 - 862 of human TRPC7.
- Tested applications:IHC, ICC/IF
- Purification:Protein G purification
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [N64A/36] antibody to TRP 7 for WB, IHC, ICC/IF, IP and antibody microarray with samples derived from human, mouse and rat.
- Validated applications: WB, IHC, ICC/IF, IP, Antibody microarray
- Recommended dilutions: WB: 1:1000, IHC: 1:1000, ICC/IF: 1:100
Type: Primary
Antigen: TRP 7
Clonality: Monoclonal
Clone: N64A/36
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat