Anti-HSP90 alpha Mouse Monoclonal Antibody [Clone: M10E3R]
Catalog # ANTIA304896-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:EL52
- Antigen symbol:HSP90 alpha
- Clonality:Monoclonal
- Clone:M10E3R
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P07900
- Antigen synonyms:Heat shock 86 kDa|heat shock 90kD protein, alpha-like 4|epididymis luminal secretory protein 52|heat shock 90kD protein 1, alpha|HS90A_HUMAN|Heat shock 90kDa protein 1 alpha|Heat shock protein HSP 90-alpha|Heat shock 90kD protein 1, alpha like 4|Heat shock protein 90kDa alpha (cytosolic) class A member 1
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 50% glycerol and 0,09% sodium azide
- Molecular weight:90 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Purified Human HSP90.
- Purification:Protein A purification
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [M10E3R] antibody to HSP90 alpha for WB and ELISA with samples derived from human.
- Validated applications: WB, ELISA
- Recommended dilutions: WB: 1:1000
Type: Primary
Antigen: HSP90 alpha
Clonality: Monoclonal
Clone: M10E3R
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human