Order Entry
Export
ContactUsLinkComponent
 
Anti-MEK1 Rabbit Polyclonal Antibody
  ANTIA304880-100
New Product
undefined
Anti-MEK1 Rabbit Polyclonal Antibody
  ANTIA304880-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    Dual specificity mitogen activated protein kinase kinase 1
  • Antigen symbol:
    MEK1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q02750
  • Antigen synonyms:
    MAPKK 1|Dual specificity mitogen-activated protein kinase kinase 1|MAP2K1|MAPKK1|MAP kinase/Erk kinase 1|MEK 1|ERK activator kinase 1|MAP kinase kinase 1|MAPK/ERK kinase 1
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,4, with 50% glycerol and 0,09% sodium azide
  • Molecular weight:
    43,4 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide from the mid - protein of Human MEK1 (aa. 250 - 350).
  • Tested applications:
    IHC
  • Purification:
    Peptide affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Rabbit polyclonal antibody to MEK1 for WB and IHC with samples derived from human.

  • Validated applications: WB, IHC
  • Recommended dilutions: WB: 1:1000, IHC: 1:50

Type: Primary
Antigen: MEK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human