Anti-DKK1 Rabbit Polyclonal Antibody
Catalog # ANTIA304872-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Dickkopf 1
- Antigen symbol:DKK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# O94907
- Antigen synonyms:Dickkopf like protein 1|DKK 1|Dickkopf homolog 1 |Dickkopf WNT signaling pathway inhibitor 1|Dickkopf 1 like|Dickkopf-related protein 1|Dickkopf-1|Dickkopf 1 homolog|Dickkopf related protein 1
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 50% glycerol and 0,09% sodium azide
- Molecular weight:28 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide from the mid - protein of Human Dickkopf - related protein 1 (aa. 120 - 220).
- Purification:Peptide affinity purification
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to DKK1 for WB with samples derived from human.
- Validated applications: WB
- Recommended dilutions: WB: 1:1000
Type: Primary
Antigen: DKK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human