Order Entry
Export
ContactUsLinkComponent
Anti-HSC70 Mouse Monoclonal Antibody [Clone: 1F2-H5]
Anti-HSC70 Mouse Monoclonal Antibody [Clone: 1F2-H5]
Catalog # ANTIA304778-200
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-HSC70 Mouse Monoclonal Antibody [Clone: 1F2-H5]
Catalog # ANTIA304778-200
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    2410008N15Rik
  • Antigen symbol:
    HSC70
  • Clonality:
    Monoclonal
  • Clone:
    1F2-H5
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P11142
  • Antigen synonyms:
    Epididymis luminal protein 33|Heat shock 70kDa protein 8|Heat shock cognate protein 54|Epididymis secretory sperm binding protein Li 72p|Heat shock 70kD protein 8|Heat shock 70 kDa protein 8|Heat shock 70kD protein 10|Heat shock cognate 71 kDa protein|Constitutive heat shock protein 70
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,4, with 50% glycerol and 0,09% sodium azide
  • Molecular weight:
    ~73 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Full length human HSC70.
  • Tested applications:
    IHC, ICC/IF
  • Purification:
    Protein G purification
  • Pack type:
    Plastic vial
  • Pk:
    200 µG

Specifications

About this item

Mouse monoclonal [1F2-H5] antibody to HSC70 for WB, IHC, ICC/IF, IP, ELISA, PLA, PBA and antibody microarray with samples derived from human, mouse and rat.

  • Validated applications: WB, IHC, ICC/IF, IP, ELISA, PLA, PBA, Antibody microarray
  • Recommended dilutions: WB: 1:1000, ICC/IF: 1:100

Type: Primary
Antigen: HSC70
Clonality: Monoclonal
Clone: 1F2-H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse, Rat