Anti-HSP27 Mouse Monoclonal Antibody [Clone: 5D12-A12]
Catalog # ANTIA304734-200
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:28 kDa heat shock protein
- Antigen symbol:HSP27
- Clonality:Monoclonal
- Clone:5D12-A12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P04792
- Antigen synonyms:epididymis secretory protein Li 102|Heat shock 27 kDa protein|Estrogen regulated 24 kDa protein|Heat shock 27kD protein 1|Heat shock 27kDa protein|Estrogen-regulated 24 kDa protein|DKFZp586P1322|CMT2F|Heat shock 25kDa protein 1
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 50% glycerol and 0,09% sodium azide
- Molecular weight:~27 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Full length human HSP27.
- Tested applications:IHC, ICC/IF
- Purification:Protein G purification
- Pack type:Plastic vial
- Pk:200 µG
Specifications
About this item
Mouse monoclonal [5D12-A12] antibody to HSP27 for WB, IHC, ICC/IF, IP and ELISA with samples derived from human.
- Validated applications: WB, IHC, ICC/IF, IP, ELISA
- Recommended dilutions: WB: 1:2000, ICC/IF: 1:100
Type: Primary
Antigen: HSP27
Clonality: Monoclonal
Clone: 5D12-A12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human