Anti-cAMP Rabbit Recombinant Antibody [Clone: RM467]
Catalog # ANTIA302470-50
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:18 kDa cationic antimicrobial protein
- Antigen symbol:cAMP
- Clonality:Recombinant
- Clone:RM467
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Form:Liquid
- Antigen synonyms:CAP 18|Adenosine 3’ 5’ cyclic monophosphate|CAP-18|cAMP|3'-5'-cyclic adenosine monophosphate|Antibacterial protein FALL-39|3' 5' cyclic adenosine monophosphate|CAMP_HUMAN|Antibacterial peptide LL-37
- Storage buffer:Supplied in phosphate buffered saline with 50% glycerol, 1% BSA, and 0,09% sodium azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:KLH - conjugated cGMP.
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:50 µG
Specifications
About this item
Rabbit monoclonal [RM467] antibody to cAMP for ELISA.
- Validated applications: ELISA
- Recommended dilutions: ELISA: 0,05 to 0,5 µg/ml
Type: Primary
Antigen: cAMP
Clonality: Recombinant
Clone: RM467
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: