Order Entry
Export
ContactUsLinkComponent
Anti-Chicken IgY Fc Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-Chicken IgY Fc Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # ANTIA296076-1
Supplier:  Antibodies.com
New Product
Anti-Chicken IgY Fc Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # ANTIA296076-1
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Secondary
  • Antigen symbol:
    Chicken IgY Fc
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Chicken
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antibody fragment:
    Whole Molecule
  • Storage buffer:
    Supplied in phosphate buffered saline with 0,2% BSA and 0,05% Pro-Clean 400
  • Storage temperature:
    Shipped at 4 °C. Store at 4 °C. Do not freeze!
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Tested applications:
    IHC, ICC
  • Purification:
    Antigen affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    1 mg

Specifications

About this item

Goat anti-chicken IgY Fc (HRP) secondary antibody for WB, IHC, ICC and ELISA with samples derived from chicken.

  • Validated applications: WB, IHC, ICC, ELISA
  • Recommended dilutions: ELISA: 1:10000 to 1:100000, ICC: 1:200 to 1:5000, IHC: 1:200 to 1:5000, WB: 1:1000 to 1:30000

Type: Secondary
Antigen: Chicken IgY Fc
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Chicken