Anti-Rabbit IgM Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
ANTIA294909-500
New Product
- Antibody type:Secondary
- Antigen symbol:Rabbit IgM
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rabbit
- Form:Liquid
- Antibody fragment:Whole Molecule
- Storage buffer:Supplied in phosphate buffered saline with 0,2% BSA and 0,09% sodium azide
- Storage temperature:Store in the dark at 4 °C. Avoid prolonged exposure to light. Do not freeze!
- Concentration:0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:IHC, ICC
- Purification:Antigen affinity purification
- Pack type:Plastic vial
- Pk:500 µG
Goat anti-rabbit IgM (FITC) secondary antibody for IHC, ICC, Flow cytometry and IF with samples derived from rabbit.
- Validated applications: IHC, ICC, Flow cytometry, IF
- Recommended dilutions: IHC: 1:50 to 1:500, ICC: 1:50 to 1:500, Flow cytometry: 1:50 to 1:200, IF: 1:50 to 1:500
Type: Secondary
Antigen: Rabbit IgM
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit