Anti-Rabbit IgM Goat Polyclonal Antibody (AP (Alkaline Phosphatase) Then)
Catalog # ANTIA294903-500
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Secondary
- Antigen symbol:Rabbit IgM
- Clonality:Polyclonal
- Conjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rabbit
- Western blot:Yes
- Form:Liquid
- Antibody fragment:Whole Molecule
- Storage buffer:Supplied in 50 mM HEPES, pH 7,1, with 0,1 M sodium chloride, 1 mM magnesium chloride, 0,1 mM zinc chloride, 0,2% BSA, and 0,09% sodium azide
- Storage temperature:Store in the dark at 4 °C. Avoid prolonged exposure to light. Do not freeze!
- Concentration:0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:IHC, ICC
- Purification:Antigen affinity purification
- Pack type:Plastic vial
- Pk:500 µG
Specifications
About this item
Goat anti-rabbit IgM (AP) secondary antibody for WB, IHC, ICC and ELISA with samples derived from rabbit.
- Validated applications: WB, IHC, ICC, ELISA
- Recommended dilutions: WB: 1:2500 to 1:25000, IHC: 1:100 to 1:1000, ICC: 1:100 to 1:1000, ELISA: 1:2500 to 1:25000
Type: Secondary
Antigen: Rabbit IgM
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit