Order Entry
Export
ContactUsLinkComponent
Anti-Rat IgM Goat Polyclonal Antibody (AP (Alkaline Phosphatase) Then)
Anti-Rat IgM Goat Polyclonal Antibody (AP (Alkaline Phosphatase) Then)
Catalog # ANTIA294701-500
Supplier:  Antibodies.com
New Product
Anti-Rat IgM Goat Polyclonal Antibody (AP (Alkaline Phosphatase) Then)
Catalog # ANTIA294701-500
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Secondary
  • Antigen symbol:
    Rat IgM
  • Clonality:
    Polyclonal
  • Conjugation:
    AP (Alkaline Phosphatase)
  • ELISA:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antibody fragment:
    Whole Molecule
  • Storage buffer:
    Supplied in 50 mM HEPES, pH 7,1, with 0,1 M sodium chloride, 1 mM magnesium chloride, 0,1 mM zinc chloride, 0,2% BSA, and 0,09% sodium azide
  • Storage temperature:
    Store in the dark at 4 °C. Avoid prolonged exposure to light. Do not freeze!
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Tested applications:
    IHC, ICC
  • Purification:
    Antigen affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    500 µG

Specifications

About this item

Goat anti-rat IgM (AP) secondary antibody for WB, IHC, ICC and ELISA with samples derived from rat.

  • Validated applications: WB, IHC, ICC, ELISA
  • Recommended dilutions: ELISA: 1:2500 to 1:25000, ICC: 1:100 to 1:1000, IHC: 1:100 to 1:1000, WB: 1:2500 to 1:25000

Type: Secondary
Antigen: Rat IgM
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat