Order Entry
Export
ContactUsLinkComponent
Anti-eIF4E (phospho Ser209) Rabbit Antibody [CLone: RM452]
Anti-eIF4E (phospho Ser209) Rabbit Antibody [CLone: RM452]
Catalog # ANTIA290832-100
Supplier:  Antibodies.com
New Product
Anti-eIF4E (phospho Ser209) Rabbit Antibody [CLone: RM452]
Catalog # ANTIA290832-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    AUTS19
  • Antigen symbol:
    eIF4E
  • Clonality:
    Recombinant
  • Clone:
    RM452
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P06730
  • Antigen synonyms:
    eIF-4F 25 kDa subunit|EIF4EL1|EIF4E1|eIF-4E|CBP|eIF 4F 25 kDa subunit|eIF4E|eIF 4E|EIF 4F
  • Modification:
    phospho Ser209
  • Storage buffer:
    Supplied in phosphate buffered saline with 50% glycerol, 1% BSA, and 0,09% sodium azide
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic phospho - peptide corresponding to human eIF4E (phospho Ser209).
  • Purification:
    Protein A chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Recombinant rabbit monoclonal [RM452] antibody to eIF4E (phospho Ser209) for WB with samples derived from human.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:1000 to 1:2000

Type: Primary
Antigen: eIF4E
Clonality: Recombinant
Clone: RM452
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human