Anti-NAT8L Goat Polyclonal Antibody
Catalog # ANTIA286080-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:1110038O08Rik
- Antigen symbol:NAT8L
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:339983, 269642, 289727
- Antigen synonyms:NAA synthetase|Hcml3|N acetyltransferase 8 like (GCN5 related, putative)|Camello-like protein 3|NAT8L|CML3|N-acetyltransferase 8-like protein|N-acetylaspartate synthetase|NAT8 like
- Storage buffer:Supplied in tris buffered saline, pH 7,3, with 0,02% sodium azide and 0,5% BSA
- Sequence:C-SVDSRFRGKGIAK
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to the internal region of mouse NAT8L.
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Goat polyclonal antibody to NAT8L for ELISA, WB, IF and Flow cytometry with samples derived from human and mouse.
- Validated applications: ELISA, WB, IF, Flow cytometry
Type: Primary
Antigen: NAT8L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse