Anti-NOTCH4 Goat Polyclonal Antibody
Catalog # ANTIA286056-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:FLJ16302
- Antigen symbol:NOTCH4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Form:Liquid
- Gene ID:4855, 18132, 406162
- Antigen synonyms:INT3|Neurogenic locus notch homolog protein 4|Notch homolog 4|NOTC4_HUMAN|NOTCH3|Notch 4 intracellular domain|hNotch4|hNotch 4|NOTCH4 protein
- Storage buffer:Supplied in tris buffered saline, pH 7,3, with 0,02% sodium azide and 0,5% BSA
- Sequence:DHFHNGHCEKGCNN
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to the internal region of human NOTCH4.
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Goat polyclonal antibody to NOTCH4 for ELISA and IF with samples derived from human and mouse.
- Validated applications: ELISA, IF
Type: Primary
Antigen: NOTCH4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse