Order Entry
Export
ContactUsLinkComponent
Anti-CD88 Goat Polyclonal Antibody
Anti-CD88 Goat Polyclonal Antibody
Catalog # ANTIA286040-100
Supplier:  Antibodies.com
New Product
Anti-CD88 Goat Polyclonal Antibody
Catalog # ANTIA286040-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ANAPHYLATOXIN RECEPTOR
  • Antigen symbol:
    CD88
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Goat
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    728, 113959
  • Antigen synonyms:
    C5a anaphylatoxin chemotactic receptor|C5a-R|c5ar1|C5aR|C5R1|C5a anaphylatoxin receptor|C5A|C5a ligand|C5AR_HUMAN
  • Storage buffer:
    Supplied in tris buffered saline, pH 7,3, with 0,02% sodium azide and 0,5% BSA
  • Sequence:
    C-RESKSFTRSTVDT
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    100 µg at 0,5 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide corresponding to the C terminus of human CD88.
  • Purification:
    Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Goat polyclonal antibody to CD88 for ELISA, WB, IF and Flow cytometry with samples derived from human.

  • Validated applications: ELISA, WB, IF, Flow cytometry

Type: Primary
Antigen: CD88
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human