Anti-COL4A6 Goat Polyclonal Antibody
ANTIA286021-100
New Product
- Antibody type:Primary
- Antigen name:COL4A6
- Antigen symbol:COL4A6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:1288
- Antigen synonyms:Collagen alpha 6(IV) chain|Collagen IV alpha 6 polypeptide|MGC88184|dJ889N15.4|Collagen type IV alpha 6|Collagen of basement membrane alpha 6|Collagen Alpha 6(IV)|Type IV alpha 6 collagen|Collagen IV alpha 6
- Storage buffer:Supplied in tris buffered saline, pH 7,3, with 0,02% sodium azide and 0,5% BSA
- Sequence:C-PSPEFETETLHNKES
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to the internal region of human COL4A6.
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide
- Pack type:Plastic vial
- Pk:100 µG
Goat polyclonal antibody to COL4A6 for ELISA, IF and Flow cytometry with samples derived from human.
- Validated applications: ELISA, IF, Flow cytometry
Type: Primary
Antigen: COL4A6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human