Order Entry
Export
ContactUsLinkComponent
Anti-HLA C Mouse Monoclonal Antibody [Clone: DT-9]
Anti-HLA C Mouse Monoclonal Antibody [Clone: DT-9]
Catalog # ANTIA285864-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-HLA C Mouse Monoclonal Antibody [Clone: DT-9]
Catalog # ANTIA285864-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    HLA C
  • Clonality:
    Monoclonal
  • Clone:
    DT-9
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Non-human primates,
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P10321
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,4, with 15 mM sodium azide
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Purified MHC class I molecules of tamarin origin.
  • Purification:
    Protein A Chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [DT-9] antibody to HLA C for IP and Flow cytometry with samples derived from non-human primates and human.

  • Validated applications: IP, Flow cytometry
  • Recommended dilutions: Flow cytometry: 1 to 4 µg/ml

Type: Primary
Antigen: HLA C
Clonality: Monoclonal
Clone: DT-9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Non-Human Primates, Human