Anti-PODXL Mouse Monoclonal Antibody [Clone: PcMab-47]
Catalog # ANTIA285849-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:GCTM-2 antigen
- Antigen symbol:PODXL
- Clonality:Monoclonal
- Clone:PcMab-47
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# O00592
- Antigen synonyms:PCLP|Podocalyxin|PCLP-1|MGC138240|PCLP1|Gp200|PC|Gp2|Pcx
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 15 mM sodium azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant PODXL ectodomain.
- Tested applications:IHC
- Purification:Protein A Chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [PcMab-47] antibody to PODXL for WB, Flow cytometry and IHC with samples derived from human.
- Validated applications: WB, Flow cytometry, IHC
- Recommended dilutions: Flow cytometry: 1 to 4 µg/ml
Type: Primary
Antigen: PODXL
Clonality: Monoclonal
Clone: PcMab-47
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human