Anti-Galectin 3 Rat Monoclonal Antibody [Clone: M3/38]
Catalog # ANTIA285837-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:35 kDa lectin
- Antigen symbol:Galectin 3
- Clonality:Monoclonal
- Clone:M3/38
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P16110
- Antigen synonyms:Carbohydrate-binding protein 35|Galactose-specific lectin 3|GAL3|CBP35|GALBP|CBP 35|Carbohydrate binding protein 35|Gal-3|Galactoside-binding protein
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 15 mM sodium azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Murine galectin - 3.
- Tested applications:IHC-P, ICC
- Purification:Protein A Chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rat monoclonal [M3/38] antibody to Galectin 3 for WB, IHC-P, ICC, Flow cytometry and IP with samples derived from human and mouse.
- Validated applications: WB, IHC-P, ICC, Flow cytometry, IP
- Recommended dilutions: Flow cytometry: 2 to 6 µg/ml
Type: Primary
Antigen: Galectin 3
Clonality: Monoclonal
Clone: M3/38
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Human, Mouse