Order Entry
Export
ContactUsLinkComponent
Anti-ACE2 Rabbit Recombinant Antibody [Clone: ACE2/6788R]
Anti-ACE2 Rabbit Recombinant Antibody [Clone: ACE2/6788R]
Catalog # ANTIA278644-100
Supplier:  Antibodies.com
New Product
undefined
Anti-ACE2 Rabbit Recombinant Antibody [Clone: ACE2/6788R]
Catalog # ANTIA278644-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ACE 2
  • Antigen symbol:
    ACE2
  • Clonality:
    Recombinant
  • Clone:
    ACE2/6788R
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q9BYF1
  • Antigen synonyms:
    ACEH|Angiotensin converting enzyme homolog|Angiotensin Converting Enzyme 2|ACE2|Angiotensin I Converting Enzyme (peptidyl dipeptidase A) 2|ACE related carboxypeptidase|ACE-related carboxypeptidase|ACE2_HUMAN|Angiotensin converting enzyme like protein
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
  • Molecular weight:
    92 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment, around amino acids 705 - 805, of human ACE2. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [ACE2/6788R] antibody to ACE2 for ELISA and IHC-P with samples derived from human.

  • Validated applications: ELISA, IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: ACE2
Clonality: Recombinant
Clone: ACE2/6788R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human