Order Entry
Export
ContactUsLinkComponent
Human BPI ELISA Kit
Human BPI ELISA Kit
Catalog # ANTIA312771-96
Supplier:  Antibodies.com
New Product
CAS Number:  
undefined
Human BPI ELISA Kit
Catalog # ANTIA312771-96
Supplier:  Antibodies.com
New Product
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    BPI
  • Description:
    Human BPI ELISA kit
  • Sample type:
    Serum, plasma or other biological fluids
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    1,5 - 24 ng/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    40 μl
  • Sensitivity:
    0,049 ng/ml
  • Regulatory status:
    RUO

Specifications

About this item

Human BPI ELISA kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human BPI in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Human BPI ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human BPI in serum, plasma or other biological fluids. An antibody specific for BPI has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the BPI present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-BPI antibody, which also binds the BPI present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-BPI antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-BPI antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of BPI captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of BPI in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, positive control, negative control, HRP-conjugated detection antibody, sample diluent, substrate solution A, substrate solution B, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.