- Pk:96 Tests
- Assay duration:One step
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:Cytokeratin 16/K16
- Description:Human cytokeratin 16/K16 ELISA kit
- Sample type:Serum, plasma or other biological fluids
- Detection method:Colorimetric
- Time to Results:1 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Detection range:0,3 - 4,8 ng/ml
- Storage temperature:4 °C
- Sample volume:40 μl
- Sensitivity:0,023 ng/ml
- Regulatory status:RUO
Human Cytokeratin 16/K16 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Cytokeratin 16/K16 in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutess, with a single wash step
- Detection range: 0,3 to 4,8 ng/ml
- Sensitivity: 0,023 ng/ml
- Sample Type: Serum, plasma or other biological fluids
- Assay Precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Human Cytokeratin 16/K16 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Cytokeratin 16/K16 in serum, plasma or other biological fluids. An antibody specific for Cytokeratin 16/K16 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Cytokeratin 16/K16 present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-Cytokeratin 16/K16 antibody, which also binds the Cytokeratin 16/K16 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-Cytokeratin 16/K16 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-Cytokeratin 16/K16 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of Cytokeratin 16/K16 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Cytokeratin 16/K16 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.