- Pk:96 Tests
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Guinea pig
- Target protein:IL-12 p40
- Description:Guinea pig IL-12 p40 ELISA kit
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Cross reactivity:Guinea pig IL-12 p40 ELISA Kit exhibits high specificity and excellent specificity for the detection of guinea pig IL-12 p40. No significant cross-reactivity or interference between IL-12 p40 and analogues was observed.
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf life:Store for 6 months at 4 °C
- Detection range:15,625 - 1000 pg/ml
- Storage temperature:4 °C
- Sample volume:100 μl
- Sensitivity:9,375 pg/ml
- Regulatory status:RUO
Guinea pig IL-12 p40 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of guinea pig IL-12 p40 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-to-use ELISA kit
- Detection Range: 15,625 to 1000 pg/ml
- Sensitivity: 9,375 pg/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Guinea Pig IL-12 p40 ELISA Kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of guinea pig IL-12 p40 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for IL-12 p40 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the IL-12 p40 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-IL-12 p40 Antibody, which binds the captured IL-12 p40 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of IL-12 p40 captured in each well. The concentration of IL-12 p40 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.