Order Entry
Export
ContactUsLinkComponent
Mouse IVL ELISA Kit
Mouse IVL ELISA Kit
Catalog # ANTIA314105-96
Supplier:  Antibodies.com
New Product
CAS Number:  
undefined
Mouse IVL ELISA Kit
Catalog # ANTIA314105-96
Supplier:  Antibodies.com
New Product
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    IVL
  • Description:
    Mouse IVL ELISA kit
  • Sample type:
    Serum, plasma or other biological fluids
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    0,75 - 12 ng/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    40 μl
  • Sensitivity:
    0,022 ng/ml
  • Regulatory status:
    RUO

Specifications

About this item

Mouse IVL ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse IVL in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection Range: 0,75 to 12 ng/mL
  • Sensitivity: 0,022 ng/ml
  • Sample Type: Serum, plasma or other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Mouse IVL ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse IVL in serum, plasma or other biological fluids. An antibody specific for IVL has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the IVL present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-IVL antibody, which also binds the IVL present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-IVL antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-IVL antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. colour develops in proportion to the amount of IVL captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of IVL in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.