Order Entry
Export
ContactUsLinkComponent
Human Angiotensin II ELISA Kit
Human Angiotensin II ELISA Kit
Catalog # ANTIA74653-96
Supplier:  Antibodies.com
New Product
CAS Number:  
undefined
Human Angiotensin II ELISA Kit
Catalog # ANTIA74653-96
Supplier:  Antibodies.com
New Product
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    Multiple steps
  • Assay Type:
    Competitive
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    angiotensin II
  • Description:
    Human angiotensin II ELISA kit
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross reactivity:
    Human angiotensin II ELISA Kit exhibits high specificity and excellent specificity for the detection of human angiotensin II. No significant cross-reactivity or interference between angiotensin II and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    31,25 - 2000 pg/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    50 μl
  • Sensitivity:
    18,75 pg/ml
  • Regulatory status:
    RUO

Specifications

About this item

Human angiotensin II ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of human angiotensin II in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Detection range: 31,25 to 2000 pg/ml
  • Sensitivity: 18,75 pg/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra to Assay: CV <8%, Inter to Assay: CV <10%

Human Angiotensin II ELISA Kit (A74653) employs the competitive enzyme immunoassay technique for the quantitative measurement of human Angiotensin II in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Angiotensin II antigen. During the incubation, Angiotensin II present in the samples or standards competes with the fixed amount of immobilised Angiotensin II for binding sites on the Biotinylated Anti-Angiotensin II Antibody. The more Angiotensin II present in a sample or standard, the less Biotinylated Anti-Angiotensin II Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Angiotensin II Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Angiotensin II present in each sample or standard. The concentration of Angiotensin II can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.