Specifications
- Pk:96 Tests
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Other
- Target protein:Apolipoprotein B
- Description:Porcine apolipoprotein B ELISA kit
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Cross reactivity:Porcine Apolipoprotein B ELISA kit exhibits high specificity and excellent specificity for the detection of porcine Apolipoprotein B. No significant cross-reactivity or interference between Apolipoprotein B and analogues was observed.
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf life:Store for 6 months at 4 °C
- Detection range:0,625 - 40 µg/ml
- Storage temperature:4 °C
- Sample volume:100 μl
- Sensitivity:0,375 µg/ml
- Regulatory status:RUO
Specifications
About this item
Porcine Apolipoprotein B ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of porcine Apolipoprotein B in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-to-use ELISA kit
- Detection range: 0,625 to 40 µg/ml
- Sensitivity: 0,375 µg/ml
- Sample type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Porcine Apolipoprotein B ELISA kit (A74661) employs the sandwich enzyme immunoassay technique for the quantitative measurement of porcine Apolipoprotein B in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Apolipoprotein B has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Apolipoprotein B present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Apolipoprotein B Antibody, which binds the captured Apolipoprotein B present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Apolipoprotein B captured in each well. The concentration of Apolipoprotein B can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.