Order Entry
Export
ContactUsLinkComponent
Human Apelin 36 ELISA Kit
Human Apelin 36 ELISA Kit
Catalog # ANTIA75200-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
undefined
Human Apelin 36 ELISA Kit
Catalog # ANTIA75200-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    Multiple steps
  • Assay Type:
    Competitive
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    Apelin 36
  • Description:
    Human apelin 36 ELISA kit
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross reactivity:
    Human Apelin 36 ELISA kit exhibits high specificity and excellent specificity for the detection of human Apelin 36. No significant cross-reactivity or interference between Apelin 36 and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    46,875 - 3000 pg/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    50 μl
  • Sensitivity:
    28,125 pg/ml
  • Regulatory status:
    RUO

Specifications

About this item

Human Apelin 36 ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of human Apelin 36 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Detection range: 46,875 to 3000 pg/ml
  • Sensitivity: 28,125 pg/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Human Apelin 36 ELISA kit (A75200) employs the competitive enzyme immunoassay technique for the quantitative measurement of human Apelin 36 in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Apelin 36 antigen. During the incubation, Apelin 36 present in the samples or standards competes with the fixed amount of immobilised Apelin 36 for binding sites on the Biotinylated Anti-Apelin 36 Antibody. The more Apelin 36 present in a sample or standard, the less Biotinylated Anti-Apelin 36 Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Apelin 36 Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Apelin 36 present in each sample or standard. The concentration of Apelin 36 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.