Specifications
- Pk:96 Tests
- Assay duration:One step
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:Cornulin
- Description:Human cornulin ELISA kit
- Sample type:Serum, plasma or other biological fluids
- Detection method:Colorimetric
- Time to Results:1 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Detection range:50 - 800 ng/L
- Storage temperature:4 °C
- Sample volume:40 μl
- Sensitivity:4,16 ng/L
- Regulatory status:RUO
Specifications
About this item
Human Cornulin ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Cornulin in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutess, with a single wash step
- Detection range: 50 to 800 ng/L
- Sensitivity: 4,16 ng/L
- Sample type: Serum, plasma or other biological fluids
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Human Cornulin ELISA kit (A313050) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Cornulin in serum, plasma or other biological fluids. An antibody specific for Cornulin has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Cornulin present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-Cornulin antibody, which also binds the Cornulin present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-Cornulin antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-Cornulin antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of Cornulin captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Cornulin in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.