- Pk:96 Tests
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:Proteasome 20S alpha 5/PSMA5
- Description:Human proteasome 20S alpha 5/PSMA5 ELISA kit
- Sample type:Serum, plasma, tissue homogenates, and other biological fluids
- Cross reactivity:Human Proteasome 20S alpha 5/PSMA5 ELISA Kit exhibits high specificity and excellent specificity for the detection of human Proteasome 20S alpha 5/PSMA5. No significant cross-reactivity or interference between Proteasome 20S alpha 5/PSMA5 and analogues was observed.
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Detection range:0,156 - 10 ng/ml
- Storage temperature:4 °C
- Sensitivity:0,094 ng/ml
- Regulatory status:RUO
Human Proteasome 20S alpha 5/PSMA5 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Proteasome 20S alpha 5/PSMA5 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready to use ELISA kit
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Human Proteasome 20S alpha 5/PSMA5 ELISA kit (A303056) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Proteasome 20S alpha 5/PSMA5 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Proteasome 20S alpha 5/PSMA5 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Proteasome 20S alpha 5/PSMA5 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Proteasome 20S alpha 5/PSMA5 Antibody, which binds the captured Proteasome 20S alpha 5/PSMA5 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Proteasome 20S alpha 5/PSMA5 captured in each well. The concentration of Proteasome 20S alpha 5/PSMA5 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.