Order Entry
Export
ContactUsLinkComponent
Human 10-HETE ELISA Kit
Human 10-HETE ELISA Kit
Catalog # ANTIA302981-96
Supplier:  Antibodies.com
New Product
CAS Number:  
undefined
Human 10-HETE ELISA Kit
Catalog # ANTIA302981-96
Supplier:  Antibodies.com
New Product
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    Multiple steps
  • Assay Type:
    Competitive
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    10-HETE
  • Description:
    Human 10-HETE ELISA kit
  • Sample type:
    Serum, plasma, tissue homogenates, and other biological fluids
  • Cross reactivity:
    Human 10-HETE ELISA Kit exhibits high specificity and excellent specificity for the detection of human 10-HETE. No significant cross-reactivity or interference between 10-HETE and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    0,156 - 10 ng/ml
  • Storage temperature:
    4 °C
  • Sensitivity:
    0,094 ng/ml
  • Regulatory status:
    RUO

Specifications

About this item

Human 10-HETE ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of human 10-HETE in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Human 10-HETE ELISA kit (A302981) employs the competitive enzyme immunoassay technique for the quantitative measurement of human 10-HETE in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with 10-HETE antigen. During the incubation, 10-HETE present in the samples or standards competes with the fixed amount of immobilised 10-HETE for binding sites on the Biotinylated anti-10-HETE antibody. The more 10-HETE present in a sample or standard, the less Biotinylated anti-10-HETE antibody that binds to the plate. Following incubation, unbound Biotinylated anti-10-HETE antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of 10-HETE present in each sample or standard. The concentration of 10-HETE can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.