Order Entry
Export
ContactUsLinkComponent
Mouse FGF-15 ELISA Kit
Mouse FGF-15 ELISA Kit
Catalog # ANTIA76538-96
Supplier:  Antibodies.com
New Product
CAS Number:  
undefined
Mouse FGF-15 ELISA Kit
Catalog # ANTIA76538-96
Supplier:  Antibodies.com
New Product
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    FGF-15
  • Description:
    Mouse FGF-15 ELISA kit
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross reactivity:
    Mouse FGF-15 ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse FGF-15. No significant cross-reactivity or interference between FGF-15 and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    4,688 - 300 pg/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    100 μl
  • Sensitivity:
    2,813 pg/ml
  • Regulatory status:
    RUO

Specifications

About this item

Mouse FGF-15 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse FGF-15 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 4,688 to 300 pg/ml
  • Sensitivity: 2,813 pg/ml
  • Sample type: Serum, plasma, tissue homogenates and other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Mouse FGF-15 ELISA kit (A76538) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse FGF-15 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for FGF-15 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the FGF-15 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-FGF-15 Antibody, which binds the captured FGF-15 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of FGF-15 captured in each well. The concentration of FGF-15 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.