Specifications
- Pk:96 Tests
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:beta Amyloid 40
- Description:Human beta amyloid 40 ELISA kit
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Cross reactivity:Human beta Amyloid 40 ELISA Kit exhibits high specificity and excellent specificity for the detection of human beta Amyloid 40. No significant cross-reactivity or interference between beta Amyloid 40 and analogues was observed.
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf life:Store for 6 months at 4 °C
- Detection range:7,813 - 500 pg/ml
- Storage temperature:4 °C
- Sample volume:100 μl
- Sensitivity:4,688 pg/ml
- Regulatory status:RUO
Specifications
About this item
Human beta Amyloid 40 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human beta Amyloid 40 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready to use ELISA kit
- Detection range: 7,813 to 500 pg/ml
- Sensitivity: 4,688 pg/ml
- Sample type: Serum, plasma, tissue homogenates and other biological fluids
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Human beta Amyloid 40 ELISA kit (A76184) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human beta Amyloid 40 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for beta Amyloid 40 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the beta Amyloid 40 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-beta Amyloid 40 Antibody, which binds the captured beta Amyloid 40 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of beta Amyloid 40 captured in each well. The concentration of beta Amyloid 40 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.