Order Entry
Export
ContactUsLinkComponent
Coleman® C300 Xtreme Gas Cartridge
Coleman® C300 Xtreme Gas Cartridge
  CAMP2182911
undefined
Coleman® C300 Xtreme Gas Cartridge
  CAMP2182911

 

  • Description:
    Coleman® C300 Xtreme gas cartridge
  • Pk:
    1

 

 

Coleman® C300 Xtreme gas cartridge with safety valve.

  • High performance blend will continue to function down to –27 °C
  • Capacity: 230 g
  • Dimensions: 9Ø×10,5H cm
  • Weight: 351 g

Equipped with a high security, self-sealing valve, this gas cartridge provides reliable performance at extreme temperatures. The cartridge be disconnected and reconnected even when it is not empty.