Order Entry
Export
ContactUsLinkComponent
 
 
p-Anisaldehyde ≥98% (by GC)
  SIAL8223141000
 :  Merck
New Product
 :  
 
p-Anisaldehyde ≥98% (by GC)
  SIAL8223141000
 :  Merck
New Product
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

  • Assay (GC, area%):
    ≥98.0% (a/a)
  • Density(d 20 °C/4 °C):
    1.121 - 1.124
  • Identity (IR):
    Passes test

 

 

 :
 :247...249 °C/1013 hPa
 :1.12 g/cm³ (25 °C)
 :M-Clarity™ quality level = MQ200
 :116 °C
 :136.15 g/mol
 :GHS08
 :Room Temperature
 :123-11-5
 :C₈H₈O₂
 :MFCD00003385