Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
AzoDye-3 CPG
AzoDye-3 CPG 1000 A (500 A)
AzoDye-3 CPG 1000 A (500 A)
AzoDye-3 CPG 1000 A (500 A)
Catalog # AATB2478
Supplier:
AAT Bioquest
New Product
CAS Number:
Expand image
AzoDye-3 CPG 1000 A (500 A)
Catalog # AATB2478
Supplier:
AAT Bioquest
New Product
CAS Number:
Specifications
Pk:
100 mg
Seq:
2440
Specifications
About this item
Product Details
:
Storage Temperature:
Freezer
Product Details