Order Entry
Export
ContactUsLinkComponent
AzoDye-2 CPG 500 A (1000 A)
AzoDye-2 CPG 500 A (1000 A)
Catalog # AATB2430
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
AzoDye-2 CPG 500 A (1000 A)
Catalog # AATB2430
Supplier:  AAT BIOQUEST
New Product
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
Storage Temperature:Freezer