Order Entry
Export
ContactUsLinkComponent
AzoDye-1 azide
AzoDye-1 azide
Catalog # AATB2412
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
AzoDye-1 azide
Catalog # AATB2412
Supplier:  AAT BIOQUEST
New Product
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
Storage Temperature:Freezer
 :572.63 g/mol