Order Entry
Export
ContactUsLinkComponent
Anti-HLA-DR Mouse Monoclonal Antibody [clone: LN3] (FITC)
Anti-HLA-DR Mouse Monoclonal Antibody [clone: LN3] (FITC)
Certificates
undefined
Anti-HLA-DR Mouse Monoclonal Antibody [clone: LN3] (FITC)

Specifications

  • Catalog No:
  • STMC60164FI
  • STMC60164FI.1
  • Antigen name:
  • HLA-DR
  • HLA-DR
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • LN3
  • LN3
  • Reactivity:
  • Rhesus monkey, Human
  • Rhesus monkey, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 3123
  • 3123
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa

Specifications