Order Entry
Export
ContactUsLinkComponent
 
Anti-Igg2b Isotype Control Mouse Monoclonal Antibody [clone: MPC-11]
 
undefined
Anti-Igg2b Isotype Control Mouse Monoclonal Antibody [clone: MPC-11]

 

  • Catalog No:
  • STMC60072
  • STMC60072.1
  • Antigen name:
  • Igg2b Isotype Control
  • Igg2b Isotype Control
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • MPC-11
  • MPC-11
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa