About this item
Rat monoclonal IgG2b antibody against human, mouse, rhesus CD11b, PerCP-Cy5.5-conjugated.
- Granulocytes
- Subsets
- Monocytes
The M1/70 antibody reacts with CD11b, an ~170 kDa type 1 transmembrane glycoprotein which associates non-covalently with CD18 to form the heterodimeric Mac-1 receptor. Through its interactions with ligands such as ICAM-1 (CD54), ICAM-2 (CD102), ICAM-4 (CD242), iC3b, and fibrinogen, Mac-1 functions in several processes, including the adherence of neutrophils and monocytes to stimulated endothelium and phagocytosis of complement-coated particles. CD11b is expressed on the surface of granulocytes, monocytes, NK cells, dendritic cells, tissue macrophages, and subsets of T and B cells, and has been used as a marker to distinguish naïve and memory CD8+ T cells. CD11b is a relatively late marker for myeloid differentiation and is undetectable on most myelomonocytic hematopoietic progenitor cells and more primitive cells. The M1/70 antibody reportedly blocks iC3b binding to Mac-1. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse Monocyte Isolation Kit (Catalog #19861).
Type:
Antigen: CD11b
Clonality: Monoclonal
Clone: M1/70
Conjugation: PerCP-Cy5.5
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Baboon, Chimpanzee, Human, Rabbit, Rhesus monkey
Specifications
- Catalog No:
- STMC60001PS.1
- STMC60001PS
- Antigen name:
- CD11b
- CD11b
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
- PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
- Clone:
- M1/70
- M1/70
- Reactivity:
- Baboon, Rabbit, Rhesus monkey, Chimpanzee, Human
- Baboon, Rabbit, Rhesus monkey, Chimpanzee, Human
- Host:
- Rat
- Rat
- Gene ID:
- 16409
- 16409
- Isotype:
- IgG2b kappa
- IgG2b kappa