Order Entry
Export
ContactUsLinkComponent
Anti-CD80 (B7-1) Hamster Monoclonal Antibody [clone: 16-10A1] (PE)
Anti-CD80 (B7-1) Hamster Monoclonal Antibody [clone: 16-10A1] (PE)
Catalog # STMC100-1622
undefined
Anti-CD80 (B7-1) Hamster Monoclonal Antibody [clone: 16-10A1] (PE)
Catalog # STMC100-1622

Specifications

  • Antigen name:
    CD80 (B7-1)
  • Clonality:
    Monoclonal
  • Clone:
    16-10A1
  • Conjugation:
    PE (Phycoerythrin)
  • Host:
    Hamster
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Gene ID:
    12519
  • Pk:
    100 µG

Specifications

About this item

Hamster (Armenian) monoclonal antibody against mouse CD80 (B7-1), PE-conjugated.

  • B cells
  • Dendritic cells
  • Macrophages
  • T cells

This monoclonal antibody reacts with mouse cluster of differentiation 80 (CD80), a 55 kDa type I transmembrane protein ligand that is part of the immunoglobulin superfamily. CD80 is expressed by macrophages, dendritic cells, and activated B cells. CD80 is closely related to and works in tandem with CD86 (B7-2) to prime T cells. It has high affinity for binding to two T cell surface antigens, CD28 and CD152 (CTLA-4), and can deliver an inhibitory signal to T cells. Use this recombinant monoclonal antibody in applications requiring detection of CD80, such as flow cytometry (FC). CD80 is significantly involved in immune cell activation in response to pathogens and acts as a cellular attachment receptor for adenovirus subgroup B. It is thought that CD80 interacts with a ligand on natural killer cells thus activating the natural killer cell-mediated cell death of the CD80 carrier. This phenomenon has potential as a possible cancer immunotherapy through the induction of CD80 expression on tumor cells. This antibody can be used as a marker to assess classically activated M1 murine macrophages.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD80 (B7-1)
Clonality: Monoclonal
Clone: 16-10A1
Conjugation: PE
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse