Order Entry
Export
ContactUsLinkComponent
Anti-CD235ab Mouse Monoclonal Antibody [clone: HIR2 (GA-R2)]
Anti-CD235ab Mouse Monoclonal Antibody [clone: HIR2 (GA-R2)]
Certificates
undefined
Anti-CD235ab Mouse Monoclonal Antibody [clone: HIR2 (GA-R2)]

Specifications

  • Catalog No:
  • STMC60111
  • STMC60111.1
  • Antigen symbol:
  • CD235ab
  • CD235ab
  • Antigen name:
  • Glycophorin A/B
  • Glycophorin A/B
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • HIR2 (GA-R2)
  • HIR2 (GA-R2)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 2993/2994
  • 2993/2994
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa

Specifications